Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Benzyl acetate

Glass bottle, 250 mL
Catalog # ROTH6197.1
Marketsource
Supplier:   Roth Carl
Benzyl acetate
Catalog # ROTH6197.1
Supplier:  Roth Carl
Marketsource
CAS Number:  140-11-4
Benzyl acetate

Glass bottle, 250 mL

CAS Number:  140-11-4
Price
Inquire For Price
Log in for availability
1 * 250 mL
Product Details & Documents
  • CAS Number:140-11-4
  • Formula:C₉H₁₀O₂
  • MDL Number:MFCD00008712
  • Molecular Weight:150.18 g/mol
  • Merck Index:13,01125
  • Boiling Pt:206 °C (1013 hPa)
  • Melting Pt:–51 °C
  • Density:1.04 g/cm³ (20 °C)
  • Flash Pt:95 °C
  • Storage Temperature:Ambient
  • EINECS:205-399-7
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.