
(2-Chlorophenyl)acetic acid methyl ester 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:57486-68-7
:
Formula:(C₆H₄Cl)CH₂COOCH₃
MDL Number:MFCD00137445
:184.62 g/mol
EINECS:260-764-8
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5240 to 1.5260 (20°C, 589 nm)
- GC:≥97.5 %