Biochemistry analyser
Catalog # YSIL2900D
Supplier: YSI LIFE SCIENCE
Specifications
- Measuring range:Glucose: 0,05 – 25 g/l ; Lactate: 0,05 – 2,70 g/l ; Glutamate: 15 – 1460 mg/l ; Glutamine: 30 – 1169 mg/l ; Glycerol: 0,75-40 g/l ; Xylose: 0,5-30 g/l ; Choline: 5 – 450 mg/l ; Hydrogen peroxide: 3 – 300 mg/l ; Sucrose: 0,1 – 25 g/l ; Ethanol: 0,04 – 3,2 g/l ; Ethanol-HC: 0,5 – 40 g/l ; Methanol: 0,1 – 2,5 g/l ; Lactose: 0,05 – 25 g/l ; Galactose: 0,1 – 25 g/l
- Memory:Yes
- Power:100 − 240 V AC/50 − 60 Hz
- Description:Biochemistry analyzer
- Display:Graphic
- Weight:12,7 kg
- Pk:1
Specifications
About this item
Biochemistry analyser features an intuitive graphical user interface, a USB port for data retrieval to measure samples from sample holders including 96 well plates and microcentrifuge tubes.
- Analyse specific results in a minute or less
- Proprietary immobilised enzyme electrodes
- Unique fluidics resist clogging and automated sample handling
Measures the chemistries in application areas such as glucose, sucrose, lactate, galactose, glutamine, lactose, glutamate, choline, xylose, glycerol, ethanol, methanol, hydrogen peroxide.
Certifications: ETL, RoHS