Order Entry
Finland
ContactUsLinkComponent
Plug set for wide mouth jerricans
Plug set for wide mouth jerricans
Catalog # HUENET-818000
Supplier:  hünersdorff GmbH
undefined
Plug set for wide mouth jerricans
Catalog # HUENET-818000
Supplier:  hünersdorff GmbH

Specifications

  • Pk:
    10

Specifications

About this item

Wide-mouth plug set for wide-mouth jerrycans.

  • Key width = 22 mm
  • Foodsafe

Set consists of: Screw plugs HDPE, natural, with wide thread 22 mm and O-ring 12×3 mm, black and O-ring 25×2 mm LDPE, blue.