0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Fast blue BB salt
Fast blue BB salt for microscopy, for histology, Sigma-Aldrich®
Fast blue BB salt for microscopy, for histology, Sigma-Aldrich®
:
SIGMA ALDRICH MICROSCOPY
Fast blue BB salt for microscopy, for histology, Sigma-Aldrich®
:
SIGMA ALDRICH MICROSCOPY
:
5486-84-0
:
:
C₃₄H₃₆Cl₄N₆O₆Zn
:
MFCD00074765
:
831.89 g/mol
:
226-817-4
:
GHS07
Merck The Color of Tissue Diagnostics