
3-Methoxy-3-methyl-1-butanol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56539-66-3
:
Formula:C₆H₁₄O₂
MDL Number:MFCD00044771
:118.18 g/mol
Boiling Pt:174 °C (760 mmHg)
Melting Pt:–50 °C
Density:0.926 g/cm³
Flash Pt:71 °C
EINECS:260-252-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4260 to 1.4280 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together