ESD safety shoes, lace-up, jo_Fit Lady Low ESD, Jori
Supplier: ELTEN
Certificates
About this item
Sporty ladies safety shoes made from nubuck leather combined with mesh inserts, skin-friendly textile lining, a full padded bellows tongue, collar padding, heel loop, and a dual-density rubber/EVA jo_SPORTY sole (oil, fuel and chemical resistant, temperature resistant from approx. -20 and +200 °C).
- Climate regulating, breathable textile lining
- Replaceable insole; orthopaedic insoles possible
- Heel pocket lining made from sturdy abrasion resistant microfibre lining
- Full-length inlay sole
- Protective steel toecap (200 J)
- Metal-free, puncture resistant midsole
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects. The shoe is tailored to the ergonomics of female feet.
Colour: Black/pink
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC
Specifications
- Catalog No:
- ELTN14121/38
- ELTN14121/37
- ELTN14121/36
- ELTN14121/42
- ELTN14121/41
- ELTN14121/40
- ELTN14121/39
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001