Bis(2-ethoxyethyl)ether for synthesis, Sigma-Aldrich®
Catalog # 8.02932.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-44 °C
- Molar Mass:162.22 g/mol
- Boiling Point:189 °C (1013 hPa)
- Vapor Pressure:0.5 hPa (20 °C)
- Flash Point:71 °C
- Refractive Index:1.4115 (20 °C, 589 nm)
- Density:0.91 g/cm3 (20 °C)
- Ignition Point:174 °C
Specification Test Results
About this item
:
:162.23 g/mol
Merck Index:13,03145
Boiling Pt:189 °C (1013 hPa)
Melting Pt:–44 °C
Density:0.91 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:203-963-7
CAS Number:112-36-7
Formula:C₈H₁₈O₃
MDL Number:MFCD00009254