Bis(2-butoxyethyl)ether for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:112-73-2
:
Formula:C₁₂H₂₆O₃
MDL Number:MFCD00009459
:218.34 g/mol
Boiling Pt:256 °C (1013 hPa)
Melting Pt:–60 °C
Density:0.88 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:204-001-9
Specification Test Results
- Solubility:3 g/l (20 °C)
- Melting Point:-60 °C
- Molar Mass:218.33 g/mol
- Boiling Point:254 °C (1013 hPa)
- Vapor Pressure:0.01 hPa (20 °C)
- Flash Point:118 °C
- Density:0.88 g/cm3 (20 °C)
- Ignition Point:190 °C