


1,2-Dimethoxyethane ≥99.9% inhibitor-free, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # 307432-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.85 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.03 %
- Peroxides (as H₂O₂):Max. 15 ppm
- Absorbance at 400 nm:Max. 0.01
- Absorbance at 350 nm:Max. 0.01
- Absorbance at 300 nm:Max. 0.03
- Absorbance at 250 nm:Max. 0.20
- Absorbance at 220 nm:Max. 1.0
- Vapor Density:3.1 (20 °C, vs air)
- Refractive Index:n20/D 1.379 (lit.)
- Explosion Limit:10.4 %
Specification Test Results
About this item
:
:90.12 g/mol
Merck Index:13,03251
Boiling Pt:85 °C (1.013 hPa)
Melting Pt:-58 °C
Melting Pt:–58 °C
Density:0.87 g/cm³ (20 °C)
Flash Pt:-2 °C
Flash Pt:–2 °C
EINECS:203-794-9
UN:2252
ADR:3,II
:GHS07
:GHS08
:GHS02
Storage Temperature:Ambient
CAS Number:110-71-4
Formula:C₄H₁₀O₂
MDL Number:MFCD00008502