Specifications
- Orbit:3 mm
- Shaking motion:Orbital
- Description:Light duty orbital shaker for microplates, SHLDMP03DG
- Model:SHLDMP03DG
- Electrical:230 V, 5,0 A, 50/60 Hz
- Tray dimensions:279×197 mm
- Max. load:4 microplates or 2 micro-tube racks
- Power:25 W
- Speed:100 - 1200 min⁻¹
- Speed accuracy:±2 %
- Timer:1 s – 160 h
- Weight:10,9 kg
- W×D×H:413×254×102 mm
- Pk:1
Specifications
About this item
This light duty microplate shaker is ideal for immunoassays and general microplate shaking, and is designed to be used in a variety of environmental conditions. Ideal for ELISA assays and DNA studies.
- Takes up less space and easily fits into most hoods and incubators
- Variable speed microprocessor control provides consistent uniform shaking action and will display last set-point and will restart if power is interrupted
- Speed ramping feature slowly increases speed to desired set-point to avoid splashing
- Spill-resistant design ensures fluids are channelled away from internal components
- Audible and visual signals will activate when system detects an obstruction or tray overload
- In timed mode, alarm will sound when the time reaches zero
- Can be used at −10 to 60 °C (max. 80% relative humidity, non condensing), in an incubator, CO₂ incubator or cold room
Touch pad controls with easy-to-read, independent LED displays for speed and time allow operator to view both settings at once. Provides repeatable and accurate results every time and is easily visible across laboratory benches. Timer will display elapsed time or, when programmed to user defined limit, will shut off unit when time reaches zero. Display will show last used settings, even after power has been turned off.
Triple eccentric drive: Permanently lubricated ball bearings and maintenance free, brushless DC motor provide reliable service and continuous duty operation.