Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Ethanolamine, Supelco®

Glass bottle, 5 mL
Catalog # 00278-5ML-F
Corrosive
Irritant
Supplier:   Merck
Ethanolamine, Supelco®
Catalog # 00278-5ML-F
Supplier:  Merck
CAS Number:  141-43-5
Ethanolamine, Supelco®
Ethanolamine, Supelco®

Glass bottle, 5 mL

CAS Number:  141-43-5
Price
127,00 €
Log in for availability
1 * 5 mL
Product Details & Documents
Synonyms:  2-Aminoethanol, Monoethanolamine
  • CAS Number:141-43-5
  • Formula:NH₂CH₂CH₂OH
  • MDL Number:MFCD00008183
  • Molecular Weight:61.08 g/mol
  • Merck Index:13,03762
  • Boiling Pt:171 °C (1013 hPa)
  • Melting Pt:10.5 °C
  • Density:1.0164 g/cm³ (20 °C)
  • Flash Pt:93 °C
  • Storage Temperature:Ambient
  • UN:2491
  • ADR:8,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:205-483-3

 

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL

Specifications