
1-Aminobutan-2-ol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13552-21-1
:
Formula:C₄H₁₁NO
MDL Number:MFCD00059196
:89.14 g/mol
Boiling Pt:169°C
UN:2735
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together