1-Propyl isothiocyanate
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:628-30-8
:
Formula:C₄H₇NS
MDL Number:MFCD00004823
:101.17 g/mol
Boiling Pt:152…153 °C (1013 hPa)
Density:0,9909 g/cm³ (0 °C)
EINECS:211-035-8
UN:2924
ADR:3,II