Glycine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:56-40-6
:
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131
:75.07 g/mol
Merck Index:13,04504
Boiling Pt:181 °C (1013 hPa)
Melting Pt:83 °C
Density:1.64 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-272-2
Specification Test Results
- Solubility:225 g/l (20 °C)
- Melting Point:232 - 236 °C (decomposition)
- Molar Mass:75.06 g/mol
- Bulk Density:920 kg/m3
- Density:1.595 g/cm3 (20 °C)
- pH:5.9 - 6.4 (50 g/l, H2O, 20 °C)