Order Entry
Finland
ContactUsLinkComponent
Glycine ≥99.5% (dried basis), Ultrapure Bioreagent for Electrophoresis, Liquid Chromatography and Molecular Biology, for liquid chromatography, J.T.Baker®
Glycine ≥99.5% (dried basis), Ultrapure Bioreagent for Electrophoresis, Liquid Chromatography and Molecular Biology, for liquid chromatography, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Glycine ≥99.5% (dried basis), Ultrapure Bioreagent for Electrophoresis, Liquid Chromatography and Molecular Biology, for liquid chromatography, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥99.5%
  • Assay (C₂H₅NO₂) (dried basis, by non-aqueous acid- base titration):
    ≥99.5 %
  • Homogeneity by TLC:
    Passes Test
  • Residue after Ignition:
    ≤0.10 %
  • Loss on Drying at 105°C:
    ≤0.2 %
  • Readily Carbonizable Substances:
    Passes Test
  • Absorbance of a 1M Solution - 280 nm:
    ≤0.100
  • RNase Activity (None Detected):
    Passes Test
  • DNase Activity (None Detected):
    Passes Test
  • Protease Activity (None Detected):
    Passes Test
  • Arsenic (As):
    ≤3 ppm
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Iron (Fe):
    ≤0.003 %

Specification Test Results

Related Information