Glycine ≥98.5% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-40-6
:
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131
:75.07 g/mol
Merck Index:14,4491
Boiling Pt:181 °C (1013 hPa)
Melting Pt:∼ 245 °C (dec.)
Density:1.595
Storage Temperature:Ambient
EINECS:200-272-2