Glycine 99+% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-40-6
:
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131
:75.07 g/mol
Merck Index:13,04504
Boiling Pt:181 °C (1013 hPa)
Melting Pt:240 °C
Density:1.595 g/cm³
Storage Temperature:Ambient
EINECS:200-272-2
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.2 % (105°C, 2 h)
- Sulfated ash:≤0.1 %
- Titration with HClO4:≥99.0 % (on dried substance)
- Chloride (Cl):≤40 ppm
- Sulfate (SO4):≤50 ppm
- Ammonium (NH4):≤200 ppm
- Arsenic (As):≤1 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
- Hydrolyzable substance:Clear solution
- Residual solvents:Meets the requirements