Sodium butyrate, Millipore®
Catalog # 567430-250
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:110.09 g/mol
Melting Pt:244 °C
Density:1.324 g/cm³ (30 °C)
EINECS:205-857-6
CAS Number:156-54-7
Formula:C₄H₇NaO₂
MDL Number:MFCD00002816