Order Entry
Finland
Orders LinkContactUsLinkComponent
Behenic acid

Behenic acid

Supplier:   Cayman Chemical
Behenic acid
Supplier:  Cayman Chemical
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  Docosanoic acid
  • CAS Number:112-85-6
  • Formula:C₂₂H₄₄O₂
  • MDL Number:MFCD00002807
  • Molecular Weight:340.59 g/mol
  • Merck Index:13,01024
  • Boiling Pt:306 °C (1013 hPa)
  • Melting Pt:80 °C
  • Density:0.8221 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-010-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 28,00 €
    28,00 €
    1 * 500 mg
       28,00 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 46,00 €
    46,00 €
    1 * 1 g
       46,00 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 81,00 €
    81,00 €
    1 * 5 g
       81,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.