Order Entry
Finland
ContactUsLinkComponent
CYGREEN® Nucleic acid Dye
CYGREEN® Nucleic acid Dye
Catalog # ENZOENZGEN1050100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
CYGREEN® Nucleic acid Dye
Catalog # ENZOENZGEN1050100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Description:
    CYGREEN® Nucleic Acid Dye
  • Seq:
    02

Specifications

About this item

Nucleic acid dye for gel staining and higher sensitivity qPCR.

  • Higher sensitivity allows detection of low target concentration
  • Greater PCR efficiency for accurate and reproducible results
  • Soluble in water for higher stability
  • Compatible with various enzyme systems
  • Suitable for commonly used rotor-type and plate-type qPCR platforms
  • Validated for qPCR applications and gel staining
  • Lower cost per assay compared to commonly used dyes

CYGREEN® Nucleic acid dye is a DNA intercalating agent that is used to stain DNA. The DNA-dye complex emits a fluorescence spectra that makes it suitable for qPCR and gel staining applications. CYGREEN® Nucleic acid dye has excitation/emission spectra similar to commonly used qPCR dyes and can be substituted in to increase sensitivity and reduce cost.