Order Entry
Finland
ContactUsLinkComponent
Urea 99.5% for molecular biology DNase-, RNase-, protease-free
Urea 99.5% for molecular biology DNase-, RNase-, protease-free
Certificates
undefined
Urea 99.5% for molecular biology DNase-, RNase-, protease-free

Specification Test Results

  • Appearance:
    White Solid
  • Assay:
    ≥99.5 %
  • UV:
    at 260 nm A: ≤0.05
  • Dnase,Rnase,Protease activity:
    none detected
  • Chloride (Cl):
    ≤5 ppm
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (as Pb):
    ≤10 ppm

Specification Test Results

Related Information