Order Entry
Finland
Orders LinkContactUsLinkComponent
Formamide

Formamide

Glass bottle for solids, 500 g
Catalog # AMBI9342
Health
Marketsource
Supplier:   Ambion
Formamide
Catalog # AMBI9342
Supplier:  Ambion
Marketsource
CAS Number:  75-12-7
Formamide
Formamide

Glass bottle for solids, 500 g

CAS Number:  75-12-7
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Carbamaldehyde
  • CAS Number:75-12-7
  • Formula:HCONH₂
  • MDL Number:MFCD00007941
  • Molecular Weight:45.04 g/mol
  • Merck Index:13,04261
  • Boiling Pt:210 °C (1013 hPa)
  • Melting Pt:2 °C
  • Density:1.1334 g/cm³ (20 °C)
  • Flash Pt:79 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-842-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.