
Formamide 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-12-7
:
Formula:HCONH₂
MDL Number:MFCD00007941
Merck Index:13,04261
Boiling Pt:210 °C (760 mmHg)
Melting Pt:2…3 °C
Density:1.133 g/cm³
Flash Pt:175 °C
:45.04 g/mol
EINECS:200-842-0
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.03 % (Coulometric)
- GC:≥99.0 %
- UV:at 275 nm A: ≤0.5
- Color scale:≤10 APHA
Specification Test Results
Frequently Bought Together