Order Entry
Finland
Orders LinkContactUsLinkComponent
3-Octanone

3-Octanone

Glass bottle for solids, 1 g
Catalog # ROTH6817.1
Marketsource
Supplier:   Roth Carl
3-Octanone
Catalog # ROTH6817.1
Supplier:  Roth Carl
Marketsource
CAS Number:  106-68-3
3-Octanone

Glass bottle for solids, 1 g

CAS Number:  106-68-3
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:106-68-3
  • Formula:C₈H₁₆O
  • MDL Number:MFCD00009515
  • Molecular Weight:128.21 g/mol
  • Merck Index:13,03799
  • Boiling Pt:168 °C (1013 hPa)
  • Melting Pt:–18.5 °C
  • Density:0.82 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • Storage Temperature:Refrigerator
  • EINECS:203-423-0
  • UN:1224
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications