


Acetonitrile ≥99.9%, TraceSELECT™ for metal speciation analysis, Riedel-de Haën™
Catalog # 01324-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Assay (GC):Min. 99.9 %
- Water (Karl Fischer):Max. 0.01 %
- Silver (Ag):Max. 0.5 ppb
- Aluminium (Al):Max. 1 ppb
- Arsenic (As):Max. 1 ppb
- Gold (Au):Max. 0.5 ppb
- Barium (Ba):Max. 0.5 ppb
- Beryllium (Be):Max. 0.5 ppb
- Bismuth (Bi):Max. 0.5 ppb
- Calcium (Ca):Max. 20 ppb
- Cadmium (Cd):Max. 0.5 ppb
- Cerium (Ce):Max. 0.5 ppb
- Cobalt (Co):Max. 0.5 ppb
- Chromium (Cr):Max. 1 ppb
- Cesium (Cs):Max. 0.5 ppb
- Copper (Cu):Max. 1 ppb
- Iron (Fe):Max. 1 ppb
- Gallium (Ga):Max. 0.5 ppb
- Germanium (Ge):Max. 0.5 ppb
- Mercury (Hg):Max. 0.5 ppb
- Indium (In):Max. 0.5 ppb
- Iridium (Ir):Max. 0.5 ppb
- Potassium (K):Max. 10 ppb
- Lithium (Li):Max. 0.5 ppb
- Magnesium (Mg):Max. 1 ppb
- Manganese (Mn):Max. 0.5 ppb
- Molybdenum (Mo):Max. 0.5 ppb
- Sodium (Na):Max. 50 ppb
- Nickel (Ni):Max. 1 ppb
- Lead (Pb):Max. 0.5 ppb
- Palladium (Pd):Max. 0.5 ppb
- Platinum (Pt):Max. 0.5 ppb
- Rubidium (Rb):Max. 0.5 ppb
- Rhodium (Rh):Max. 0.5 ppb
- Ruthenium (Ru):Max. 0.5 ppb
- Antimony (Sb):Max. 0.5 ppb
- Selenium (Se):Max. 1 ppb
- Tin (Sn):Max. 0.5 ppb
- Strontium (Sr):Max. 0.5 ppb
- Titanium (Ti):Max. 1 ppb
- Thallium (Tl):Max. 0.5 ppb
- Vanadium (V):Max. 0.5 ppb
- Zinc (Zn):Max. 1 ppb
- Zirconium (Zr):Max. 0.5 ppb
- Bromide (Br):Max. 10 ppb
- Chloride (Cl):Max. 25 ppb
- Fluoride (F):Max. 10 ppb
- Iodide (I):Max. 10 ppb
- Nitrite (NO₂):Max. 10 ppb
- Nitrate (NO₃):Max. 25 ppb
- Phosphate (PO₄):Max. 10 ppb
- Sulfate (SO₄):Max. 10 ppb
- Transmittance at 195 nm:Min. 80 %
- Transmittance at 200 nm:Min. 95 %
- Transmittance at 230 nm:Min. 99 %
Specification Test Results
About this item
:
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:82 °C (1.013 hPa)
Melting Pt:–46 °C
Density:0.7857 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS08
:GHS07
:GHS02
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878