Order Entry
Finland
Orders LinkContactUsLinkComponent
D-(-)-Mannitol analytical reagent

D-(-)-Mannitol analytical reagent

Plastic bottle for solids, 500 g
Catalog # 25314.264
Supplier:   VWR Chemicals
D-(-)-Mannitol analytical reagent
Catalog # 25314.264
Supplier:  VWR Chemicals
CAS Number:  69-65-8
D-(-)-Mannitol analytical reagent
D-(-)-Mannitol analytical reagent

Plastic bottle for solids, 500 g

CAS Number:  69-65-8
Price
74,30 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290…295 °C (4 hPa)
  • Melting Pt:164…169 °C
  • Density:1.49 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-711-8
Specifications

Specification Test Results

  • Reducing sugars:
    Passes test
  • Acidity or alkalinity:
    Passes test
  • Melting point:
    165 - 170 °C
  • Specific optical rotation (8 %; borate):
    23 - 24 °
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue (SO4):
    Max. 0.05 %
  • Loss on drying (100°C):
    Max. 0.3 %
  • Cl (Chloride):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • As (Arsenic):
    Max. 2 ppm

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.