ESD safety shoes, clogs, Mia
: ELTEN
Lightweight anti-static clogs made from smooth, tear resistant microfibre upper, an adjustable heel strap, plus a mono-density PU sole (2,5 mm, oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
- Climate regulating, breathable textile lining
- Full length inlay sole (C-Fit ESD)
- ESD soft fleece insole
- Shock-absorbent heel
- Protective steel toecap (200 J)
Ergonomically tailored to female feet.
Colour: Black or white
: EN 61340-5-1 (ESD); EN ISO 20345 SB, SRC, A, E, FO.
- Catalog No:
- ELTN74440/42
- ELTN74430/42
- ELTN74440/39
- ELTN74440/38
- ELTN74430/41
- ELTN74440/37
- ELTN74430/40
- ELTN74430/39
- ELTN74440/36
- ELTN74440/35
- ELTN74430/38
- ELTN74430/37
- ELTN74430/36
- ELTN74430/35
- ELTN74440/41
- ELTN74440/40
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001