(S)-(+)-2-Amino-1-propanol
Catalog # MOLEM58220382
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:75.11 g/mol
Boiling Pt:173 °C (1013 hPa)
Density:0.955 g/cm³ (20 °C)
EINECS:220-388-7
:GHS05
Storage Temperature:Ambient
CAS Number:2749-11-3
Formula:C₃H₉NO
MDL Number:MFCD00064412