Order Entry
Finland
ContactUsLinkComponent
Organic reference standard, 2,3',5-Trichlorobiphenyl (PCB No. 26) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,3',5-Trichlorobiphenyl (PCB No. 26) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-26
CAS Number:  
Organic reference standard, 2,3',5-Trichlorobiphenyl (PCB No. 26) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-26
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    38444-81-4
  • Description:
    2,3',5-Trichlorobiphenyl (PCB No. 26) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₇Cl₃
  • Description:
    2,3',5-Trichlorobiphenyl (PCB No. 26)

Specifications

About this item