Order Entry
Finland
ContactUsLinkComponent
Organic reference standard, 3,3',4,4',5-Pentachlorobiphenyl (PCB No. 126) 35 µg/ml in isooctane
Organic reference standard, 3,3',4,4',5-Pentachlorobiphenyl (PCB No. 126) 35 µg/ml in isooctane
Catalog # PROOCIL-PCB-126
CAS Number:  
Organic reference standard, 3,3',4,4',5-Pentachlorobiphenyl (PCB No. 126) 35 µg/ml in isooctane
Catalog # PROOCIL-PCB-126
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    57465-28-8
  • Description:
    3,3',4,4',5-Pentachlorobiphenyl (PCB No. 126) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    3,3',4,4',5-Pentachlorobiphenyl (PCB No. 126)

Specifications

About this item