Order Entry
Finland
ContactUsLinkComponent
 
Organic reference standard, 3,3',4,4',5-Pentachlorobiphenyl (PCB No. 126) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-126-CS
 :  
Organic reference standard, 3,3',4,4',5-Pentachlorobiphenyl (PCB No. 126) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-126-CS
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    57465-28-8
  • Description:
    3,3',4,4',5-Pentachlorobiphenyl (PCB No. 126) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    3,3',4,4',5-Pentachlorobiphenyl (PCB No. 126)