Order Entry
Finland
ContactUsLinkComponent
Organic reference standard, 2,2',3,3',6-Pentachlorobiphenyl (PCB No. 84) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,2',3,3',6-Pentachlorobiphenyl (PCB No. 84) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-84
 :  
Organic reference standard, 2,2',3,3',6-Pentachlorobiphenyl (PCB No. 84) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-84
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    52663-60-2
  • Description:
    2,2',3,3',6-Pentachlorobiphenyl (PCB No. 84) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,2',3,3',6-Pentachlorobiphenyl (PCB No. 84)