Order Entry
Finland
ContactUsLinkComponent
 
Organic reference standard, 2,2',4,4',5,6'-Hexachlorobiphenyl (PCB No. 154) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-154
 :  
Organic reference standard, 2,2',4,4',5,6'-Hexachlorobiphenyl (PCB No. 154) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-154
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    60145-22-4
  • Description:
    2,2',4,4',5,6'-Hexachlorobiphenyl (PCB No. 154) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₄Cl₆
  • Description:
    2,2',4,4',5,6'-Hexachlorobiphenyl (PCB No. 154)