Order Entry
Finland
ContactUsLinkComponent
Organic reference standard, 2,2',3,3',4,5',6,6'-Octachlorobiphenyl (PCB No. 200) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,2',3,3',4,5',6,6'-Octachlorobiphenyl (PCB No. 200) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-200
CAS Number:  
Organic reference standard, 2,2',3,3',4,5',6,6'-Octachlorobiphenyl (PCB No. 200) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-200
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    40186-71-8
  • Description:
    2,2',3,3',4,5',6,6'-Octachlorobiphenyl (PCB No. 200) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₂Cl₈
  • Description:
    2,2',3,3',4,5',6,6'-Octachlorobiphenyl (PCB No. 200)

Specifications

About this item