0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Gentamicin sulphate
Gentamicin sulphate
Gentamicin sulphate
Gentamicin sulphate
Supplier:
BIOWEST
Certificates
Expand image
Gentamicin sulphate
Supplier:
BIOWEST
Certificates
About this item
CAS Number:
1405-41-0
:
MDL Number:
MFCD00270181
Related Information
Documentation
Certificates
Search by Lot Number
Documentation