Organic reference standard, 1,2-Dichloropropane 1000 µg/ml in methanol
Catalog # PROONE5627
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,5 mL
- CAS No.:78-87-5
- Description:1,2-Dichloropropane 1,000 µg/ml in methanol
- Formula:C₃H₆Cl₂
- Description:1,2-Dichloropropane