Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(-)-Mannitol
D-(-)-Mannitol, EMPROVE® EXPERT Ph. Eur., BP, USP, JP
D-(-)-Mannitol, EMPROVE® EXPERT Ph. Eur., BP, USP, JP
:
MERCK PRODUCTION CHEMICALS
D-(-)-Mannitol, EMPROVE® EXPERT Ph. Eur., BP, USP, JP
:
MERCK PRODUCTION CHEMICALS
:
69-65-8
:
:
HOH₂C(CH(OH))₄CH₂OH
:
MFCD00064287
:
182.17 g/mol
:
13,05769
:
290…295 °C (4 hPa)
:
164…169 °C
:
1.49 g/cm³ (20 °C)
:
Ambient
:
200-711-8
MERP1.37096.1000_TDS_EN