Order Entry
Finland
ContactUsLinkComponent
Ammonium heptamolybdate tetrahydrate 99+% for analysis
Irritant
Ammonium heptamolybdate tetrahydrate 99+% for analysis
Certificates
undefined
Irritant
Ammonium heptamolybdate tetrahydrate 99+% for analysis
Certificates

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Assay:
    ≥99.0 %
  • Chloride (Cl):
    ≤10 ppm
  • Phosphate (PO4):
    ≤5 ppm
  • Sulfate (SO4):
    ≤100 ppm
  • Heavy metals (as Pb):
    ≤10 ppm

Specification Test Results

Related Information

Frequently Bought Together