Order Entry
Finland
Orders LinkContactUsLinkComponent
Talc, TECHNICAL

Talc, TECHNICAL

Plastic bucket, 5 kg
Catalog # 28454.360
Supplier:   VWR Chemicals
Talc, TECHNICAL
Catalog # 28454.360
Supplier:  VWR Chemicals
CAS Number:  14807-96-6
Talc, TECHNICAL
Talc, TECHNICAL

Plastic bucket, 5 kg

CAS Number:  14807-96-6
Price
75,80 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Magnesium silicate monohydrate, Soapstone, Talcum venetum
  • CAS Number:14807-96-6
  • Formula:Mg₃O₁₁Si₄·1H₂O
  • MDL Number:MFCD00792903
  • Molecular Weight:379.27 g/mol
  • Merck Index:13,09127
  • Melting Pt:> 1200 °C
  • Density:2.58…2.83 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:238-877-9
Specifications

Specification Test Results

  • Loss on ignition:
    Max. 10 %

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.