Order Entry
Finland
ContactUsLinkComponent
Tetrasodium diphosphate decahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Irritant
Tetrasodium diphosphate decahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
Irritant
Tetrasodium diphosphate decahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    99.0 - 103.0 %
  • Chloride (Cl):
    max. 0.002 %
  • Iron (Fe):
    max. 0.001 %
  • Heavy Metals (as Pb):
    max. 0.001 %
  • Insoluble Matter:
    max. 0.01 %
  • Nitrogen Compounds (as N):
    max. 0.001 %
  • pH of 5% Solution at 25°C:
    9.5 - 10.5
  • Sulfate (SO4):
    max. 0.005 %

Specification Test Results

Related Information