Order Entry
Finland
ContactUsLinkComponent
5-Sulphosalicylic acid dihydrate ≥99.0% (by acidimetry) for synthesis, Sigma-Aldrich®
5-Sulphosalicylic acid dihydrate ≥99.0% (by acidimetry) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
5-Sulphosalicylic acid dihydrate ≥99.0% (by acidimetry) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (acidimetric):
    ≥ 99.0 %
  • Heavy metals (as Pb):
    ≤ 0.001 %
  • Identity (IR):
    passes test
  • Synonyms:
    2-Hydroxybenzoic-5-Sulfonic Acid, Dihydrate
  • Chemical Formula:
    HOC6H3(COOH)SO3H·2H2O

Specification Test Results

Related Information