5-Sulphosalicylic acid dihydrate 99+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5965-83-3
:
Formula:HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
MDL Number:MFCD00149540
:254.22 g/mol
Merck Index:14,8964
Melting Pt:103…112 °C
Density:1.695 g/cm³ (20 °C)
Flash Pt:150 °C
Storage Temperature:Ambient
EINECS:202-555-6
UN:2585
ADR:8,III
:GHS05
:GHS07