

EDTA tetrasodium salt dihydrate
Catalog # MOLE54799339-1KG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
Merck Index:13,03544
Melting Pt:> 300 °C
Flash Pt:>100 °C
:416.2 g/mol
EINECS:200-573-9
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:10378-23-1
Formula:C₁₀H₁₂N₂Na₄O₈·2H₂O
MDL Number:MFCD00150026