

Titriplex® III (ethylenedinitrilotetraacetic acid disodium salt dihydrate), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:6381-92-6
:
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037
:372.24 g/mol
Merck Index:13,03543
Melting Pt:255 °C
:M-Clarity™ quality level = MQ500 (more info.)
EINECS:205-358-3
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (calc. on dried substance):99.0- 101 .0 %
- Assay (calc. as dihydrate):99.0- 101.0 %
- Identification (IR):Complies
- ldentity complexation (edetate):Complies
- ldentity of sodium:Complies
- Appearance:White to almost white, crystalline powder
- Appearance of solution (5%, water):Clear and colorless
- pH (1%, water):4.3-4.7
- pH (5%, water):4.0- 5.5
- Cyanide (CN):Not more than 1 ppm
- Heavy metals (as Pb):Not more than 10 ppm
- Arsenic (As):Not more than 1 ppm
- Iron (Fe):Not more than 5 ppm
- Calcium (Ca):Complies
- Nitrilotriacetic acid:Not more than 0.1 %
- Methanol (GC):Not more than 3000 ppm
- Loss on drying:8. 7 - 11.4 %
- Sulphated ash:37.0- 39.0 %
- Other residual solvents (ICH Q3C):Excluded by production process
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERP1.08421.1000_TDS_EN MERP1.08421.2500_TDS_EN MERP1.08421.9025_TDS_EN ProductPage_108421
Product specification
msds_108421