Order Entry
Finland
ContactUsLinkComponent
EDTA disodium salt dihydrate
EDTA disodium salt dihydrate
Catalog # 705-0079
Supplier:  Hach
CAS Number:  
undefined
EDTA disodium salt dihydrate
Catalog # 705-0079
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :372.24 g/mol
Merck Index:13,03543
Melting Pt:255 °C
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:6381-92-6
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037