tri-Sodium citrate dihydrate ≥99%, Reagent Grade
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:6132-04-3
:
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031
:294.1 g/mol
Merck Index:13,08675
Melting Pt:150 °C
Density:1.76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-675-3
Specification Test Results
- Calcium:≤0.005%
- Chloride:≤0.005%
- DNase:none detected
- Heavy Metals (as Pb):≤0.001%
- Insolubles:≤0.005%
- Iron:≤0.0005%
- Loss on Drying (%):10.0 – 13.0
- pH (5%, Water) @25 °C:7.0 – 9.0
- Protease:none detected
- Purity (Dry Basis) (%):≥99%
- RNase:none detected
- Sulphate:≤0.005%
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_VWRC0101
SDS
VWR Chemicals Big Packs and Bulks