Order Entry
Finland
Orders LinkContactUsLinkComponent
Ammonia solution 25%

Ammonia solution 25%

25 L
Catalog # SWEDAM871302
Corrosive
Irritant
Environment
Supplier:   SWED HANDLING
Ammonia solution 25%
Catalog # SWEDAM871302
Supplier:  SWED HANDLING
CAS Number:  1336-21-6
Ammonia solution 25%

25 L

CAS Number:  1336-21-6
Price
114,00 €
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88…0.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pk:
    25 L

Specifications

Suositukset

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.