Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium pyruvate
Sodium pyruvate 0.1 mol/l in aqueous solution
Sodium pyruvate 0.1 mol/l in aqueous solution
EURLECM0542D
:
EUROCLONE
:
Sodium pyruvate 0.1 mol/l in aqueous solution
EURLECM0542D
:
EUROCLONE
:
Pack type:
Glass bottle
Pk:
100 mL
Seq:
2440
:
:
110.04 g/mol
:
Ambient
:
204-024-4
:
113-24-6
:
CH₃COCOONa
:
MFCD00002586