Order Entry
Finland
ContactUsLinkComponent
 
 
 
 
(±)-3-Chloro-1,2-propanediol 99%
 
undefined
 
 
 
(±)-3-Chloro-1,2-propanediol 99%

 

  • Appearance:
    Clear light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4790 to 1.4810 (20°C, 589 nm)
  • GC:
    ≥98.5 %